Grey Safety Goggles
Grey Safety Goggles
Actual product may vary
Grey Safety Goggles
Grey Safety Goggles
Grey Safety Goggles
Polyester headband provides a comfortable fit, while indirect venting allows air flow for anti-fog protection.
Have Questions?
Catalog NumberProduct Type
FIS19-181-513Safety/Chemical Splash Goggles
Catalog number FIS19-181-513
Price (SGD)
11.00
EA
Add to cart
Product Type:
Safety/Chemical Splash Goggles
Price (SGD)
11.00
EA
Add to cart
Streamlined design to work with hardhats and respirators.
  • Polycarbonate lens with anti-fog and anti-scratch coating
  • Frame consists of a combination of a polycarbonate lens with a Thermoplastic rubber lining
  • Provides chemical splash protection
  • Indirect venting allows airflow to minimize fogging
  • Comfortable polyester headband provides a comfortable fit
  • ANSI/ISEA Z87.1-2020: Z87+ U6 D3 D4
Specifications
Lens CoatingAnti-fog, Scratch-resistant
Certifications/ComplianceANSI/ISEA Z87.1-2020: Z87+ U6 D3 D4
Lens TintClear
Product TypeSafety/Chemical Splash Goggles
Frame ColorLight Gray
MaterialPolyester
Frame MaterialPolycarbonate, TPR
Unit SizeEA