Ultra-Utility Box™
Actual product may vary
Youngstown Barrel and Drum
Ultra-Utility Box™
Choose a Youngstown Ultra-Utility Box™ for outdoor storage, spill kits or docks.
Have Questions?
Catalog NumberColorIncludesWeight (English)
FIS19-171-084Forest Green5 in. Solid Rubber, 2 rigid and 2 swivel wheel with brake68 lb.
Catalog number FIS19-171-084
Price (SGD)
-
Color:
Forest Green
Includes:
5 in. Solid Rubber, 2 rigid and 2 swivel wheel with brake
Weight (English):
68 lb.
  • Heavy-duty double-walled lid sloped to shed rain
  • Volume: 56.8L (15 gal.)
  • 122 x 7.8cm (48 x 31 in.)
  • Forkliftable
  • Lockable
  • Rubber wheel measurement: 12.7cm (5 in.)
  • Pneumatic wheel measurement: 20.32cm (8 in.)
Specifications
Depth (English) Interior43.5 in.
Depth (Metric) Exterior122 cm
Depth (Metric) Interior110.4 cm
Dimensions (D x W x H)48 x 31 x 38 in.
For Use With (Equipment)Spill kit, storage bin and much more
Height (English) Interior25 in.
Height (Metric) Exterior96.6 cm
Height (Metric) Interior63.5 cm
Includes5 in. Solid Rubber, 2 rigid and 2 swivel wheel with brake
Load Bearing Capacity15 gal.
Weight (English)68 lb.
Weight (Metric)31 kg
Width (English) Interior24 in.
Width (Metric) Exterior78.7 cm
Width (Metric) Interior60.9 cm
ColorForest Green
Depth (English) Exterior48 in.
Height (English) Exterior38 in.
MaterialPolyethylene
Product TypeUtility Box
Width (English) Exterior31 in.
Unit SizeEach