Spilfyter™ Streefyter™ Double-Weight Universal Sorbent Pad
Actual product may vary
NPS Corp.
Spilfyter™ Streefyter™ Double-Weight Universal Sorbent Pad
Light-duty pad yet tough enough for daily use
Have Questions?
Catalog NumberCapacity (English)
FIS19-169-70228 gal.
Catalog number FIS19-169-702
Price (SGD)
181.00
Case
Capacity (English):
28 gal.
Price (SGD)
181.00
Case
Ideal for use in many maintenance and cleanup applications—use to keep floors clean and dry, place under and around machinery, use to line shelves or as a general-purpose wiper
  • Exclusive Grab and Go™ packaging allows for easy portability and storage
  • Bales stack neatly on top of one another—no more tipping pallets
  • Shrink wrapped polybag fits tight around sorbent pads—keep pads protected when in storage or when kept outside in the elements
  • Absorbs and retains oil, water, solvents and coolants
  • No perforations or zips
  • Color: gray
  • Use as a rag for quick wipe-ups
  • L x W: 18 x 16 in. (45.7 x 40.6cm)
  • Sorbing capacity: 26 gal. (98L)
Specifications
ColorGray
Capacity (English)28 gal.
Capacity (Metric)105.99 L
MaterialPolypropylene
TypeUniversal Sorbent Pad
For Use With (Application)Ideal for use in many maintenance and cleanup applications, absorbs and retains oil, water, solvents, coolants, etc
Weight CapacityHeavyweight
Dimensions (L x W)18 x 16 in. (45.72 x 40.6 cm)
Length (English)18 in.
Length (Metric)45.72 cm
Width (English)16 in.
Width (Metric)40.6 cm
Weight (English)14 lb.
Weight (Metric)6.35 kg
Unit SizeCase