G29 Chemical Gloves
Actual product may vary
G29 Chemical Gloves
KleenGuard™ G29 Neoprene Chemical Gloves with Dual-Dex™ Technology provide the dexterity and tactile sensitivity of thin mil gloves with greater durability and reduced exposure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-167-723Large, 9
FIS19-167-722Medium, 8
Catalog number FIS19-167-723
Price (SGD)
-
Size:
Large, 9
  • Neoprene blend construction designed for protection from solvents
  • Textured fingertips for wet or dry grip
  • Not made with natural rubber latex
  • Silicone-free
  • Powder free
  • Ambidextrous for greater efficiency
  • Highest level of dexterity per EN420:2003 standard
  • Available in sizes 6 (XS) - 11 (2XL)
Common Applications:
  • Handling hydraulic fluids
  • Working with solvents such as Frekote, IPA, and Varson
  • Wiping down parts and machinery with solvents
  • Janitorial cleaning
Specifications
ANSI Abrasion RatingANSI 2
Certifications/Compliance (Europe)CE 0120
ColorBlue
HandAmbidextrous
Latex FreeYes
MaterialNeoprene Blend
Product TypeNeoprene Gloves
SizeLarge, 9
Thickness (English)12 mil
Thickness (Metric)0.30 mm
EN388 StandardEN 388:4101X
EN 421No
EN ISO 374-1Type B
EN ISO 374-1 CHEMICAL LETTER CODESKOP
EN ISO 374 - 5Bacteria, Fungi WITH Virus Protection
For Use With (Application)Protection from solvents
Glove Cuff StyleBeaded/Rolled
Grip DesignDual-Dex™ Technology
LiningUnlined
Model49825
SeriesKleenGuard™ G29
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizePack