SGD-20 Safety Glasses Dispensers
Actual product may vary
SGD-20 Safety Glasses Dispensers
Holds up to 20 pair of glasses
Have Questions?
Catalog NumberDescription
FIS19-162-412With protective cover
Catalog number FIS19-162-412
Price (SGD)
-
Description:
With protective cover
  • Made of 1/8 in. (0.3cm) clear acrylic
  • Mounts to wall
  • Flame treated edges for smoothness
  • Available with lid for extra protection
Specifications
Height (English)12 in.
Height (Metric)30.5 cm
Width (English)15 in.
Width (Metric)38.1 cm
DescriptionWith protective cover
Depth (Metric)17.8 cm
Device Holds20 pairs of glasses
Depth (English)7 in.
Product TypeEyewear storage shelf
IncludesProtective cover
MaterialAcrylic
Unit SizeEach