Softies™ Disposable Ear Plugs
Actual product may vary
Moldex
Softies™ Disposable Ear Plugs
Softies™ disposable ear plugs are made of extra-soft, extra-light foam allowing a gentle seal and comfortable fit.
Have Questions?
Catalog NumberCorded
FIS19-160-0162Corded
Catalog number FIS19-160-0162
Price (SGD)
123.00
Case
Corded:
Corded
Price (SGD)
123.00
Case
  • Radiant orange color for easy compliance checks
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Shaped to fit even very small ear canals
Specifications
ANSI S3.1974Yes
Cord ColorOrange
Cord MaterialVinyl
CSA Z94.2Class AL
SizeUniversal
For Use With (Application)Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
High VisibilityYes
Packaging TypePoly bag
Pairs per Package100 Pairs/Pk.
Plug MaterialFoam
Plug ColorOrange
Noise Reduction Rating (NRR)33 dB
CordedCorded
Plug ShapeBullet
DisposableSingle-use
Metal DetectableNo
Unit SizeCase