Chemcor™ Lined Countertop Corrosives/Acids Safety Cabinet
Actual product may vary
Justrite
Chemcor™ Lined Countertop Corrosives/Acids Safety Cabinet
Smaller sized cabinets fit nearly anywhere
Have Questions?
Catalog NumberDoor Style
FIS19-159-712Manual-close
Catalog number FIS19-159-712
Price (SGD)
4,903.00
Each
Door Style:
Manual-close
Price (SGD)
4,903.00
Each
  • Thermoplastic coating on interior surface to provide protection
  • Non-porous surface resists staining
  • Double-wall, 18-gauge (1mm) welded steel construction
  • Liquid-tight containment sump 51mm (2 in.)
  • Features dual vents, adjustable shelves, three-point stainless steel bullet self-latching doors with U·Loc™ padlockable handle and Haz-Alert™ reflective warning labels
  • Manual-close door(s) opens to full 180 degrees and self-latches
Specifications
Certifications/ComplianceNFPA, OSHA
Depth (Metric) Exterior43.2 cm
Description18-gauge double-walled steel
Dimensions (D x W x H) Interior13.75 x 13.75 x 18.5 in.
Door StyleManual-close
For Use With (Application)Flammables
Height (Metric) Exterior55.9 cm
IncludesChemCor™ liner, welded Steel double wall construction, 2 in. liquid tight containment sump, dual vents, adjustable shelves, three-point Stainless Steel bullet self latching doors with U-Loc™ padlockable handle, kickplat
Load Bearing Capacity4 gal. (15 L)
No. of Doors1
No. of Shelves1
Surface CoatingEpoxy
Weight (English)63 lb.
Weight (Metric)29 kg
Width (Metric) Exterior43.2 cm
ColorBlue
Depth (English) Exterior17 in.
Height (English) Exterior22 in.
MaterialSteel
Product TypeChemCor™ Lined Countertop Safety Cabinet
Width (English) Exterior17 in.
Unit SizeEach