Cleanroom Adhesive Mats
Actual product may vary
Valutek
Cleanroom Adhesive Mats
Effectively capture dirt and dust from foot-traffic and equipment wheels before they enter a controlled environment
Have Questions?
Catalog NumberColorDimensions (L x W)
FIS19-150-1195Blue18 x 36 in.
Catalog number FIS19-150-1195
Price (SGD)
165.00
Case
Color:
Blue
Dimensions (L x W):
18 x 36 in.
Price (SGD)
165.00
Case
  • Constructed of 1.9 mil / 50 micron LDPE (Low Density Polyethylene)
  • Multi-layered clear-film full adhesive layers
  • Adhesive is a 0.03 mil / 8 micron water-based acrylic
  • Adhesive force: 650/800g/25mm
  • Tensile strength: 170 kg/cm2
  • Rupture elongation: >250%
  • Heat resistance: 70°C (158°F) - 48 hours
  • 30 layers
Peel Adhesion:
  • Individual layers: 6.5 oz. / 180g / 25mm ±20
  • Bottom Layer: 25 oz. / 700g /25mm ±50
Tab Properties:
  • Tab Size: 1 × 1 in.
  • Tab Color: White
Specifications
Dimensions (L x W)18 x 36 in.
For Use With (Application)Capture dirt and dust from foot-traffic and equipment wheels before they enter the controlled environment
Quantity30 Sheets/Mat
ColorBlue
Unit SizeCase