MaxiChem™ 14 in. Nitrile Coated Chemical Resistant Gloves
Actual product may vary
PIP
MaxiChem™ 14 in. Nitrile Coated Chemical Resistant Gloves
MaxiChem™ gloves are designed for those working with chemicals requiring longer and higher levels of protection. Nitrile synthetic coating combined with a light seamless knit liner provides comfort, sensitivity, fit and dexterity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-148-1802X-Large
FIS19-148-176Small
FIS19-148-178Large
FIS19-148-179X-Large
Catalog number FIS19-148-180
Price (SGD)
-
Size:
2X-Large
  • Chemical Resistance - Delivered through the ATG™ LiquiTech™ technology platform
  • Ultra Lightweight - Synthetic coating combined with a light seamless knit liner
  • Tailored Cuff - Keeping the cuff close to your hand so maintaining your fingers at the tip of the glove to maximize dexterity
  • Micro-cup non-slip grip finish allows for a controlled and secure grip in oily and wet applications
Specifications
Certifications/ComplianceISO 9001:2000, CE, ISO 14001:2004
Certifications/Compliance (Europe)CE, EN 374-3
ChlorinatedNo
ColorBlack, Green
Latex FreeYes
MaterialNitrile
Size2X-Large
Thickness (English)51 mil
Thickness (Metric)1.29 mm
EN388 StandardEN 388:4121
EN 421No
EN ISO 374-1 CHEMICAL LETTER CODESAKL
For Use With (Application)Chemical sampling, offshore Petroleum, Acids and Caustics, anodizing, Solvent based cleaning, black and green liquor processes, fertilizer and pesticide handling
Glove Cuff StyleGauntlet
Grip DesignNon-slip
Liner MaterialNylon/Lycra
LiningLined
Model56-635/XXL
SeriesMaxiChem™ 56-635
Length (English)14 in.
Length (Metric)35.5 cm
Unit SizeDozen