Chemical Yellow Absorbent Pad
Actual product may vary
Chemical Yellow Absorbent Pad
Color-coded for easy identification of different types of spills and hazards
Have Questions?
Catalog NumberCapacity (English)
FIS19-140-91822 gal.
Catalog number FIS19-140-918
Price (SGD)
171.00
Pack
Capacity (English):
22 gal.
Price (SGD)
171.00
Pack
Color-coded for easy identification of different types of spills and hazards
  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • BRIGHTSORB™ High Visibility Series feature safety signage and absorbency in one
  • Perforations provide cost control and reduce waste by allowing you to use only what you need
Specifications
ColorYellow
Capacity (English)22 gal.
Capacity (Metric)83.2 L
MaterialPolypropylene
TypeChemical Absorbent Pad
Packaging Quantity100/Cs.
Weight CapacityHeavyweight
Dimensions (L x W)48.2 x 38.1 cm (19 x 15 in.)
Length (English)19 in.
Length (Metric)48.2 cm
Width (English)15 in.
Width (Metric)38.1 cm
Unit SizePack