▸
First Aid and Medical
▸
First Aid Kits and Stations
▸
Arsenal™ Trauma Bags
Actual product may vary
Arsenal™ Trauma Bags
Portable equipment bags for emergency use
Have Questions?
Change view
Catalog Number
Color
Model
FIS19-130-4573
Blue
Arsenal GB5220
FIS19-130-4569
Blue
Arsenal GB5210
2 Options
Catalog number
FIS19-130-4573
Price (SGD)
161.00
Each
Add to cart
Color:
Blue
Model:
Arsenal GB5220
Price (SGD)
161.00
Each
Add to cart
600D polyester with PVC backing
ID card window
Multi-pocketed
Back Pack
3 in. padded shoulder straps with reflective striping
51 × 33 × 15cm (20 × 13 × 6 in.) main hold
Reflective trim on main panel
Phone hold
Carry handle
Responder
Six division multi–tiered mesh interior pocket
18 × 43 × 25cm (7 × 17 × 10 in.) main hold
20 × 3 × 20cm (8 × 1 × 8 in.) front pocket
Small Trauma
20 × 28 × 20cm (8 × 11 × 8 in.) main hold
Two 20 × 5 × 15cm (8 × 2 × 6 in.) side pockets
28 × 5 × 15cm (11 × 2 × 6 in.) front pocket
Reflective stripe
Large Trauma
31 × 36 × 23cm (12 × 14 × 9 in.) main hold
Two 31 × 8 × 13cm (12 × 3 × 5 in.) side pockets
Adjustable internal dividers
Reflective stripe
Fire fighters, EMTs, Police, Public safety
Specifications
For Use With (Application)
Fire fighters, EMTs, police, public safety
Product Type
Trauma Bag
Color
Blue
Model
Arsenal GB5220