Disposable Polypropylene Coveralls
Actual product may vary
Tians
Disposable Polypropylene Coveralls
Low lint coverall for use in ISO cleanroom class 7 critical environments and many industrial applications. Nonwoven fabric is cloth-like and cool to wear.
Have Questions?
Catalog NumberSize
FIS19-130-2073DX-Large
Catalog number FIS19-130-2073D
Price (SGD)
203.00
Case
Size:
X-Large
Price (SGD)
203.00
Case
  • Economical white polypropylene coverall is ideal low-cost alternative for general protection in nonhazardous environments
  • Cool, comfortable, and lightweight
  • Zipper front, elastic wrist and ankles, and folded collar
Specifications
Product TypeCleanroom Coveralls
SizeX-Large
GenderUnisex
MaterialPolypropylene
ColorWhite
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic
Neck StyleCollared
Closure TypeZipper
SterilityNon-sterile
For Use With (Application)Controlled Environments
Cleanroom ClassClass 10,000 (ISO 7)
Unit SizeCase