Critical Cover™ AlphaGuard™ Lab Coats
Actual product may vary
Critical Cover™ AlphaGuard™ Lab Coats
Critical Cover™ AlphaGuard™ Lab Coats
Critical Cover™ AlphaGuard™ Lab Coats
Critical Cover™ AlphaGuard™ Lab Coats
AlphaProTech
Critical Cover™ AlphaGuard™ Lab Coats
Soft and breathable multilayer SMS material provides comfort in critical environments. Choose from elastic or knit wrist styles in blue or white colors.
Have Questions?
Catalog NumberSizeColorCuff Style
FIS19-130-1466BMediumWhiteKnit
Catalog number FIS19-130-1466B
Price (SGD)
458.00
Case
Size:
Medium
Color:
White
Cuff Style:
Knit
Price (SGD)
458.00
Case
Soft and breathable multilayer SMS material provides comfort in critical environments. Choose from elastic or knit wrist styles in blue or white colors.
  • Soft, breathable and fluid repellent
  • Static dissipative
  • Ultra clean, low particle count
Specifications
Product TypeControlled Environments Lab Coat
SizeMedium
MaterialSMS
ColorWhite
Closure TypeSnaps
Cuff StyleKnit
DisposableSingle-use
SterilityNon-sterile
No. of Pockets3
Collar TypeTapered
Latex FreeYes
Seam TypeSerged
Size (English) Chest25.25 in.
Size (Metric) Chest63.5 cm
Length (English)39.5 in.
Length (Metric)100.3 cm
Length (Metric) Sleeve76.8 cm
Unit SizeCase