Critical Cover™ AlphaGuard™ Frocks
Actual product may vary
AlphaProTech
Critical Cover™ AlphaGuard™ Frocks
Frocks are constructed with low particle count multilayer SMS material to offer high levels of breathability without compromising filtration efficiency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-130-1465CLarge
FIS19-130-1465ASmall
FIS19-130-1465BMedium
Catalog number FIS19-130-1465C
Price (SGD)
472.00
Case
Size:
Large
Price (SGD)
472.00
Case
  • Low particle count
  • Fluid repellent
  • Clean, durable and soft
  • Static dissipative
  • Raglan sleeves
  • Snap collar and close
  • Serged seams
Specifications
SizeLarge
MaterialSMS
ColorWhite
SterilityNon-sterile
Closure TypeSnaps
Cuff StyleElastic
Collar TypeMandarin
Size (English) Chest23.5 in.
Size (Metric) Chest59.69 cm
Sleeve StyleRaglan Sleeve
Length (Metric) Sleeve90.17 cm
Product TypeControlled Environments Frock
Seam TypeSerged
Unit SizeCase