Search Fisher Scientific
| Catalog Number | Dimensions (L x W) | Thickness (English) |
|---|---|---|
| FIS19-130-1222 | 36 x 24 in. (91 x 61 cm) | 6 mil |
| FIS19-130-1214 | 5 x 3 in. (12.7 x 7.6 cm) | 6 mil |
| FIS19-130-1219 | 15 x 12 in. (38 x 30.5 cm) | 6 mil |
| FIS19-130-1220 | 18 x 12 in. (46 x 30.5 cm) | 6 mil |
| FIS19-130-1221 | 24 x 18 in. (61 x 46 cm) | 6 mil |
| FIS19-121-180 | 10 x 8 in. (25.4 x 20 cm) | 4 mil |
| FIS19-130-0645 | 24 x 20 in. (61 x 50.8 cm) | 4 mil |