Clear Polyethylene Cleanroom Bags
Clear Polyethylene Cleanroom Bags
Actual product may vary
KNF FLEXPAK
Clear Polyethylene Cleanroom Bags
Ultraclean polybags can safely store parts and/or materials that require a moderate level (Level II and Level III) of packaging cleanliness
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Thickness (English)
FIS19-130-121915 x 12 in. (38 x 30.5 cm)6 mil
FIS19-130-12145 x 3 in. (12.7 x 7.6 cm)6 mil
FIS19-130-122018 x 12 in. (46 x 30.5 cm)6 mil
FIS19-130-122124 x 18 in. (61 x 46 cm)6 mil
FIS19-130-122236 x 24 in. (91 x 61 cm)6 mil
FIS19-121-18010 x 8 in. (25.4 x 20 cm)4 mil
FIS19-130-064524 x 20 in. (61 x 50.8 cm)4 mil
Catalog number FIS19-130-1219
Price (SGD)
51.00
PK
Add to cart
Dimensions (L x W):
15 x 12 in. (38 x 30.5 cm)
Thickness (English):
6 mil
Price (SGD)
51.00
PK
Add to cart
Ultraclean polybags can safely store parts and/or materials that require a moderate level (Level II and Level III) of packaging cleanliness
  • Polyethylene is an ideal material for general-purpose packaging; has a tensile strength of 2100psi, and a melting point of 107°C (225°F)
  • Approved for cleanroom use in pharmaceuticals, aerospace, semiconductors, electronics, food packaging and many other medical or mechanical applications
  • Antistatic bags are amine-free; ideal for general-purpose packaging applications requiring antistatic properties
Order Info:  Vacuum sealed and packaged in a Class 100 cleanroom. Available in 2, 3, 4 and 6mil thickness in several sizes. Contact your Fisher Scientific Safety Customer Service Representative for the complete listing of sizes and ordering information.
Specifications
AutoclavableNot Autoclavable
Certifications/ComplianceFDA Approved, Meet NASA JPG 5322.1 Level 100. A
ColorClear
DisposableSingle-use
MaterialPolyethylene
Temperature Range (Metric)107°C
Thickness (English)6 mil
Dimensions (L x W)15 x 12 in. (38 x 30.5 cm)
Length (English)15 in.
Length (Metric)38 cm
Width (English)12 in.
Width (Metric)30.5 cm
Product TypeCleanroom Bags
Unit SizePK