Paper Heat-Seal Pouches
Paper Heat-Seal Pouches
Actual product may vary
Cardinal Health
Paper Heat-Seal Pouches
With blue film
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)
FIS19-130-004513 x 7.5 in. (33 x 19cm)
FIS19-130-004410 x 6 in. (25.4 x 15.2cm)
FIS19-130-004922 x 18 in. (55.9 x 45.7cm)
Catalog number FIS19-130-0045
Price (SGD)
469.00
CS
Add to cart
Dimensions (L x W):
13 x 7.5 in. (33 x 19cm)
Price (SGD)
469.00
CS
Add to cart
  • Fracture-resistant film with high-strength surgical-grade paper provide adequate breathability and required sterile barrier
  • Peelable, low-lint design
  • Dual chemical indicators visually identify packages exposed to steam or EtO sterilization
Specifications
AutoclavableAutoclavable
Closure TypeHeat Seal
For Use With (Application)Designed for quick, easy presentation of sterile items, can be used in steam or Ethylene Oxide (EO) sterilization
IncludesBlue film
MaterialPaper
Dimensions (L x W)13 x 7.5 in. (33 x 19cm)
Length (English)13 in.
Length (Metric)33 cm
Width (English)7.5 in.
Width (Metric)19 cm
Unit SizeCS