Paper Self-Seal Pouches
Actual product may vary
Cardinal Health
Paper Self-Seal Pouches
No need to heat seal
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-130-00409.875 x 3.5 in. (25.1 x 8.9 cm)
Catalog number FIS19-130-0040
Price (SGD)
-
Dimensions (L x W):
9.875 x 3.5 in. (25.1 x 8.9 cm)
No need to heat seal
  • Extra-strength adhesive provides heat-seal performance with self-seal convenience
  • Blue fracture-resistant film and high-strength surgical-grade paper provide adequate breathability and required sterile barrier
  • Peelable, low-lint design
  • Dual-chemical indicators visually identify packages that have been exposed to either steam or EtO sterilization
Specifications
DescriptionWith adhesive
AutoclavableAutoclavable
Closure TypeSelf-seal
ColorWhite
DisposableSingle-use
For Use With (Application)Designed for quick, easy presentation of sterile items, can be used in steam or Ethylene Oxide (EO) sterilization
IncludesBlue film
MaterialPaper
SterilityNon-sterile
Dimensions (L x W)9.875 x 3.5 in. (25.1 x 8.9 cm)
Length (English)9.875 in.
Width (English)3.5 in.
Product TypeSterilization Pouch
Length (Metric)25.1 cm
Width (Metric)8.9 cm
Unit SizeCase