UT1225 Cleanroom BOPP Tape
Actual product may vary
UltraTape
UT1225 Cleanroom BOPP Tape
Designed for cleanroom packaging and in process use where residue is not a concern. UltraTape UT1225 Cleanroom BOPP Tape has a thin backing a low stretch that make it ideal for automated equipment and dispensers. It is useful in the removal of protective films and bag sealing.
Have Questions?
Catalog NumberColor
FIS19-120-3776Clear
Catalog number FIS19-120-3776
Price (SGD)
145.00
Pack
Color:
Clear
Price (SGD)
145.00
Pack
  • Bag sealing and packaging where removal and reuse is not intended
  • Aids in the removal of protective films
  • Very low stretch
  • Operating temperature: 32 to 140°F (0 to 60°C)
  • Bonding surface should be clean and dry for best adhesion
  • Strength of adhesive bond is dependent on the amount of surface area directly in contact with the adhesive
  • Firm pressure is recommended to achieve good adhesive to surface contact
  • Store at 70°F (21°C) with 50% relative humidity
Specifications
Adhesive TypeAcrylic
ColorClear
MaterialBOPP
Product TypePackaging tape
For Use With (Application)Cleanroom packaging and process use
Tensile Strength20 lb./in.
Thickness (English)2.6 mil
Length (English)55 yd.
Unit SizePack