Search Fisher Scientific
| Catalog Number | Material | Product Type | Size |
|---|---|---|---|
| FIS19-120-2481 | Plastic Inner Core | Tacky Roller | 45.7 cm (18 in.) |
| FIS19-161-925 | Metal | Extendable Handle | 122 to 244 cm (48 to 96 in.) |
| FIS19-003-433 | Nickel-coated Metal | Roller Mop Handle | 45.7 cm (18 in.) |
| FIS19-808-718 | Plastic Inner Core Foam | Tacky Roller | 45.7 cm (18 in.) |