KleenGuard™ A40 Coveralls, Elastic Wrists and Ankles
Actual product may vary
Ansell
KleenGuard™ A40 Coveralls, Elastic Wrists and Ankles
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
Have Questions?
Catalog NumberSizeAnkle StyleNeck Style
FIS19-120-2399CLargeElasticCollared
Catalog number FIS19-120-2399C
Price (SGD)
547.00
Case
Size:
Large
Ankle Style:
Elastic
Neck Style:
Collared
Price (SGD)
547.00
Case
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
  • Breathable, microporous film laminate fabric.
  • Barrier protection against hazardous particulates and aerosols and against low-hazard liquid splash.
  • ASTM F1670 for blood penetration and ASTM F1671 for bloodborne pathogens.
  • ASTM F903 Penetration Testing provides passing results for isocyanate paint (40%) and Potassium Chromate (20%).
  • Anti-stat according to NFPA 99.
  • Features a zippered front and come in a variety of sizes.
  • Eligible for recycling via RightCycle when used in non-hazardous applications as classified by federal, state, and local regulations.
  • Elastic wrists and ankles.
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
Specifications
Product TypeGeneral Purpose Coveralls
SizeLarge
GenderUnisex
MaterialMicroporous Film Laminate
ColorWhite
DisposableLimited-use
Ankle StyleElastic
Cuff StyleElastic
Neck StyleCollared
Hood TypeUnhooded
Closure TypeFront Zipper
SterilityNon-sterile
ResistanceParticle, Liquid
Certifications/ComplianceASTM F1670, ASTM F1671, ASTM F903, NFPA 99
ModelKleenGuard™ A40
For Use With (Application)Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Maintenance and General Manufacturing
StyleElastic Wrists and Ankles
Unit SizeCase