Sure-Grip™ EX Flammable Safety Cabinet
Actual product may vary
Justrite
Sure-Grip™ EX Flammable Safety Cabinet
Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.
Have Questions?
Catalog NumberLoad Bearing CapacityDimensions (D x W x H) InteriorDoor StyleWeight (Metric)
FIS19-120-233760 gal.30.34 x 30.75 x 60.5 in.Manual-close141 kg
Catalog number FIS19-120-2337
Price (SGD)
6,200.00
Each
Load Bearing Capacity:
60 gal.
Dimensions (D x W x H) Interior:
30.34 x 30.75 x 60.5 in.
Door Style:
Manual-close
Weight (Metric):
141 kg
Price (SGD)
6,200.00
Each
Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.
  • Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
  • Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
  • Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self-latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Meet OSHA and NFPA requirements
Specifications
Certifications/ComplianceFM, NFPA, OSHA
Depth (English) Interior30.34 in.
Depth (Metric) Exterior86.4 cm
Depth (Metric) Interior77.1 cm
Dimensions (D x W x H) Interior30.34 x 30.75 x 60.5 in.
Door StyleManual-close
For Use With (Application)Flammable liquid storage and protection
Height (English) Interior60.5 in.
Height (Metric) Exterior165.1 cm
Height (Metric) Interior153.7 cm
IncludesU-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with
Load Bearing Capacity60 gal.
No. of Doors2
No. of Shelves2
Surface CoatingEpoxy, Polyester
Weight (English)310 lb.
Weight (Metric)141 kg
Width (English) Interior30.75 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior78 cm
ColorYellow
Depth (English) Exterior34 in.
Height (English) Exterior65 in.
MaterialSteel (CR)
Product TypeCan Storage Cabinet
Width (English) Exterior34 in.
Unit SizeEach