Kut-Gard™ 07-K320 Seamless Knit Aramid Fiber/Cotton Plated Gloves
Actual product may vary
PIP
Kut-Gard™ 07-K320 Seamless Knit Aramid Fiber/Cotton Plated Gloves
Cotton-plated aramid fiber construction provides heat resistance, cut protection, and is inherently flame resistant.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-100-909Large
FIS19-100-907Small
FIS19-100-908Medium
FIS19-100-910X-Large
Catalog number FIS19-100-909
Price (SGD)
188.00
Dozen
Size:
Large
Price (SGD)
188.00
Dozen
  • Aramid fiber construction provides cut protection, heat resistance, and is inherently flame resistant
  • Seamless knit design protects hands without sacrificing comfort or dexterity
  • Material will not melt, ignite, or conduct electricity
  • Ambidextrous pattern reduces replacement costs
  • Color coded hems for easy size identification
  • Can be used alone, or as glove liners
Specifications
ConstructionSeamless Knit
Coating MaterialUncoated
ColorYellow
Cut Resistance RatingANSI A1
Exterior FinishNo Texture
HandAmbidextrous
MaterialCotton, Aramid Fiber
Product TypeSeamless Knit Gloves
SizeLarge
StyleReversible
For Use With (Application)Automotive Assembly, Light Heat, Glass Operations, and Metal Stamping
Glove Cuff StyleKnit
Hem ColorYellow
Series07-K320
Yarn Gauge7
Length (English)10.5 in.
Length (Metric)26.7 cm
Unit SizeDozen