Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Fit Testing Kits and Supplies
▸
Bitrex Fit Test Kit: Solution
Actual product may vary
Allegro
Bitrex Fit Test Kit: Solution
Offers qualitative OSHA compliant fit testing for disposable and reusable dust and mist respirators.
Have Questions?
Catalog Number
Product Type
FIS19-088-510
Test Solution
Catalog number
FIS19-088-510
Price (SGD)
62.00
Pack
Add to cart
Product Type:
Test Solution
Price (SGD)
62.00
Pack
Add to cart
Bitrex Sensitivity Solution and Bitrex Test Solution
Box of 6 glass ampules each
With pre-drilled plastic cap for easy pouring
Plastic protection from broken glass
One-time use
Averages 3 to 5 fit tests per tube
Contains 2.5cc of solution
Specifications
Product Type
Test Solution
Certifications/Compliance
OSHA
For Use With (Equipment)
Allegro Bitrex Fit Test Kit (2041)
Model
2041-12K
Quantity
6/Pk.
Type (Solution)
Test solution