Banana Oil Fit-Test Kits
Actual product may vary
Allegro
Banana Oil Fit-Test Kits
Isoamyl Acetate, also referred to as Banana Oil, is a qualitative fit test agent used to detect respirator leakage.
Have Questions?
Catalog NumberIncludesProduct Type
FIS19-088-506Two 1 oz. bottles of test agents, vinyl test enclosure, measuring pipette and paper towels in a handy carry bagStandard Banana Oil Fit Test Kit
Catalog number FIS19-088-506
Price (SGD)
233.00
Each
Includes:
Two 1 oz. bottles of test agents, vinyl test enclosure, measuring pipette and paper towels in a handy carry bag
Product Type:
Standard Banana Oil Fit Test Kit
Price (SGD)
233.00
Each
  • Banana oil (isoamyl acetate) is the substance originally used for testing respirator fit; its strong, recognizable odor quickly indicates the lack of proper fit
  • Available in two fit test kits and Fit Check Ampules that provide a fast and easy method of on-the-spot qualitative fit testing
Specifications
Product TypeStandard Banana Oil Fit Test Kit
For Use With (Application)Respirator Fit Testing
IncludesTwo 1 oz. bottles of test agents, vinyl test enclosure, measuring pipette and paper towels in a handy carry bag
Model0203
Quantity1 Ea.
Unit SizeEach