Check Order Status
Belt-Mounted and Vehicle-Mounted PAPR System Accessory, Cartridges
Actual product may vary
Fisherbrand
Belt-Mounted and Vehicle-Mounted PAPR System Accessory, Cartridges
Choose from a large selection of cartridges for powered air purifying respirators used in a variety of industrial environments.
Have Questions?
Catalog Number
Cartridge Type
Color
For Use With (Application)
FIS19-087391
High Efficiency Particulate Filter (HE)
Magenta
Transportation, Mining, Oil & Gas, General Manufacturing
Catalog number
FIS19-087391
Price (SGD)
-
Cartridge Type:
High Efficiency Particulate Filter (HE)
Color:
Magenta
For Use With (Application):
Transportation, Mining, Oil & Gas, General Manufacturing
Cartridge attaches to the motor blower
Filters supply air to PAPR headgear of choice
Products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product
3M Catalog# GVP-440, GVP-441
Specifications
Cartridge Type
High Efficiency Particulate Filter (HE)
Certifications/Compliance
NIOSH
Color
Magenta
Product Type
Cartridge
For Use With (Application)
Transportation, Mining, Oil & Gas, General Manufacturing
For Use With (Equipment)
Belt-mounted and vehicle-mounted PAPR systems
Model
GVP-440
Quantity
8/Cs.