Foam Ear Plugs
Foam Ear Plugs
Actual product may vary
Fisherbrand
Foam Ear Plugs
Foam ear plugs are an economical choice for disposable hearing protection. Hypoallergenic and a tapered design help provide a noise-reducing seal in the ear canal.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelCorded
FIS19-0720761100Uncorded
FKS12580147K1110Corded
Catalog number FIS19-072076
Price (SGD)
98.00
Pack
Add to cart
Model:
1100
Corded:
Uncorded
Price (SGD)
98.00
Pack
Add to cart
  • Disposable orange ear plugs
  • Made from hypoallergenic material for maximum protection and comfort
  • Smooth, dirt-resistant surface for hygiene
  • Recommended for high noise situations (up to 100 dBA TWA)
Specifications
ANSI S3.1974Yes
CSA Z94.2Class AL
SizeUniversal
For Use With (Application)Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High VisibilityYes
Model1100
Packaging TypePoly bag
Pairs per Package1000 Pairs/Cs
Plug MaterialFoam
Plug ColorOrange
Noise Reduction Rating (NRR)29 dB
CordedUncorded
Plug ShapeBullet
DisposableSingle-use
Metal DetectableNo
Unit SizePack