Spilfyter™ Grab and Go Neutralizer Spill Kits
Actual product may vary
NPS Corp.
Spilfyter™ Grab and Go Neutralizer Spill Kits
Handle small acid or base spills quickly
Have Questions?
Catalog number FIS19-066-470
Price (SGD)
465.00
Each
Price (SGD)
465.00
Each
  • Compact spill kit with everything needed for small acid/base spills
  • KolorSafe™ liquid neutralizers have color indicator for easy identification of neutralization
  • Plastic case with handle makes the kit easy to store and carry
Specifications
TypeNeutralizer Spill Kit
Includes1 qt. liquid acid neutralizer , 1 qt. liquid base neutralizer, 2 Aualockit™ shaker bottle, 1 pair nitrile gloves, 1 pair splash goggles wisk on pan trigger sprayer disposal bag with twist tie
Weight (English)11 lb.
Weight (Metric)5 kg
Unit SizeEach