Scott™ Pro Scottfold* Towels
Actual product may vary
Ansell
Scott™ Pro Scottfold* Towels
FSC Certified Product. Larger case size to reduce packaging waste. Meets EPA standards with a minimum of 40% post-consumer recycled fiber & total min. of 50% recycled fiber. Delivers one-at-a-time dispensing every time.
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-065-79912.4 x 7.8 in.
Catalog number FIS19-065-799
Price (SGD)
221.00
Case
Dimensions (L x W):
12.4 x 7.8 in.
Price (SGD)
221.00
Case
  • 4,375 paper towels per case; 175 towels per pack; 25 packs per case
  • Scott Pro Scottfold paper towels feature a patented fold that creates 2-ply thickness and eliminates tearing and tabbing, so your guests and employees will waste less
  • The white paper disposable hand towels feature exclusive Absorbency Pockets for better absorbency
  • Hygienic, touchless dispensing with most universal folded towel dispensers: only touch the drying towel you use
  • The Scott Pro Brand guarantees a 10% reduction of cost (vs. C-fold towels)
Specifications
Product TypeHand Towel
Length (English)12.4 in.
Length (Metric)31.4 cm
MaterialPaper
Certifications/ComplianceEPA
ColorWhite
Dimensions (L x W)12.4 x 7.8 in.
For Use With (Application)For general purpose cleaning and drying
Unit SizeCase