Prefabricated Quick-Disconnect Safety Disposal Cans
Actual product may vary
Justrite
Prefabricated Quick-Disconnect Safety Disposal Cans
For continuous collection of HPLC waste flow
Have Questions?
Catalog NumberCapacity (English)Diameter (Metric) OuterHeight (Metric)Includes
FIS19-059-3792 gal.30.5 cm37.5 cmStainless Steel Fittings
Catalog number FIS19-059-379
Price (SGD)
2,668.00
Each
Capacity (English):
2 gal.
Diameter (Metric) Outer:
30.5 cm
Height (Metric):
37.5 cm
Includes:
Stainless Steel Fittings
Price (SGD)
2,668.00
Each
For continuous collection of HPLC waste flow
Polyethylene can with quick-disconnect system permits “hands-free” liquid waste accumulation for maximum protection against chemicals. Suitable for pump-fed or gravity flow HPLC waste methods.
  • Allows free flow of liquid from outside source into FM-approved polyethylene disposal can
  • Two fittings accommodate tubing with 0.25 to 0.375 in. diameter (0.6 to 1.0cm)
  • Built-in pressure-relief prevents excessive pressure in extreme temperatures
  • Translucent white color makes detection of liquid level easy
  • Available with stainless-steel fitting with PTFE O-rings or economical polypropylene fitting with EPDM O-rings
Specifications
Capacity (Metric)8 L
Certifications/ComplianceFM, OSHA, EPA
Diameter (Metric) Outer30.5 cm
Diameter (English) Outer12 in.
Capacity (English)2 gal.
ColorWhite
Height (Metric)37.5 cm
Quantity1 Ea.
Diameter (English) Opening3.65 in.
Diameter (Metric) Opening9.3 cm
IncludesStainless Steel Fittings
For Use With (Application)For continuous collection of HPLC waste flow
Height (English)14.75 in.
MaterialPolyethylene
Product TypeSafety Disposal Can
Unit SizeEach