High-Leverage Pail Opener
Actual product may vary
Youngstown Barrel and Drum
High-Leverage Pail Opener
Angled design eliminates cover distortion
Have Questions?
Catalog NumberLength (English)
FIS19-057-53016 in.
Catalog number FIS19-057-530
Price (SGD)
119.00
Each
Length (English):
16 in.
Price (SGD)
119.00
Each
  • Cast aluminum
  • Length: 40.64cm (16 in.)
  • Weight: 0.227kg (8oz)
Specifications
Product TypePail Opener
Length (English)16 in.
Length (Metric)40.64 cm
MaterialCast Aluminum
Unit SizeEach