White Nylon Hairnets
Actual product may vary
West Chester
White Nylon Hairnets
Nylon hairnets feature an elastic adjustment and is available in 18 in. or 21 in. diameter.
Have Questions?
Catalog NumberDiameter (English)Diameter (Metric)Size
FIS19-053-10018 in.45.7 cm18 in.
Catalog number FIS19-053-100
Price (SGD)
208.00
Case
Diameter (English):
18 in.
Diameter (Metric):
45.7 cm
Size:
18 in.
Price (SGD)
208.00
Case
  • Nylon hairnet
  • Ideal for use in food service, food prep, manufacturing, and retail applications
  • Elastic band for flexible fit
  • Available in 18 in. or 21 in. sizes
  • White
Specifications
Product TypeHairnet
Size18 in.
MaterialNylon
ColorWhite
DisposableSingle-use
Diameter (English)18 in.
Diameter (Metric)45.7 cm
Unit SizeCase