Mini Safety Cabinet
Mini Safety Cabinet
Actual product may vary
Mini Safety Cabinet
Mini Safety Cabinet
Mini Safety Cabinet
Justrite
Mini Safety Cabinet
For safe, transport/ storage of oils, chemicals and cleaners
Have Questions?
Catalog NumberColor
FIS19-052-392Yellow
Catalog number FIS19-052-392
Price (SGD)
2,142.00
Each
Add to cart
Color:
Yellow
Price (SGD)
2,142.00
Each
Add to cart
  • Ideal for manufacturing facilities, amusement parks, movie sets, hotels and any recreational facility using small quantities of flammables for cleaning and maintenance
  • Fire-resistant cabinet with small footprint for easy mobility on service vehicles, carts and maintenance trikes for point-of-use applications.
  • Features a leaktight 2 in. (51mm) well, flush mounted U-Loc™ handle and accepts optional padlock
  • Durable steel construction with a tough powder paint finish and three-point latching system
Specifications
Certifications/ComplianceOSHA, NFPA Code 30, UFC 79
Depth (English) Interior4.75 in.
Depth (Metric) Exterior20.3 cm
Depth (Metric) Interior12 cm
Dimensions (D x W x H) Interior4.75 x 13.77 x 12.15 in. (12 x 35 x 30.9 cm)
Door StyleManual-close
For Use With (Application)Flammables
Height (English) Interior12.15 in.
Height (Metric) Exterior55.9 cm
Height (Metric) Interior30.9 cm
IncludesTwo key, one adjustable shelf and three bungee cord, small footprint, U-Loc™ handles with two keys, Haz-Alert™ reflective trilingual exterior labeling, leakproof 2 in. bottom sump, dual vents with flame arresters, four
Load Bearing Capacity1 gal.
No. of Doors1
No. of Shelves1
Surface CoatingEpoxy, Polyester
Weight (English)43 lb.
Weight (Metric)19 kg
Width (English) Interior13.77 in.
Width (Metric) Exterior43.2 cm
Width (Metric) Interior35 cm
ColorYellow
Depth (English) Exterior8 in.
Height (English) Exterior22 in.
MaterialSteel (CR)
Product TypeSafety Cabinet
Width (English) Exterior17 in.
Unit SizeEach