Temp-Tec™ NL-517 Chemical Resistant Gloves with Thermal Insulation
Actual product may vary
Temp-Tec™ NL-517 Chemical Resistant Gloves with Thermal Insulation
Temp-Tec™ NL-517 Chemical Resistant Gloves with Thermal Insulation
MAPA
Temp-Tec™ NL-517 Chemical Resistant Gloves with Thermal Insulation
Heavyweight construction provides multi-purpose chemical resistance to acids and aliphatic solvents. The double-knit cotton liner protects hands during high temperature applications.
Have Questions?
Catalog NumberSize
FIS19-050-302C10
Catalog number FIS19-050-302C
Price (SGD)
106.00
Pair
Size:
10
Price (SGD)
106.00
Pair
Protection from extreme temperatures
  • Heavyweight construction provides multi-purpose chemical resistance to acids and aliphatic solvents
  • Double-knit cotton liner protects hands during high temperature applications
  • Neoprene coating with pebbled-finish enhances grip on wet or slippery objects
  • Extended length, 17 in. gauntlet cuff for additional arm protection
  • FDA approved, Standard 21 CFR
Specifications
Certifications/ComplianceFDA 21 CFR 170-199
Coating MaterialNeoprene
ColorBlack, Blue
Exterior FinishPartially Textured
Size10
Temperature (Metric) Max300°F
EN388 Standard2212
EN ISO 374-1 CHEMICAL LETTER CODESACJKL
For Use With (Application)Boiler Room Work, Chemical Treatment, Hot Pipe Handling, Steam Cleaning, and Polymer Extrusion
Glove Cuff StyleGauntlet
Liner MaterialDouble-Knit Cotton
SeriesTemp-Tec™ NL-517
Yarn Gauge15
Length (English)17 in.
Length (Metric)43.2 cm
Unit SizePair