HyFlex™ 11-800 Foam Nitrile Coated Work Gloves
Actual product may vary
HyFlex™ 11-800 Foam Nitrile Coated Work Gloves
HyFlex™ 11-800 Foam Nitrile Coated Work Gloves
HyFlex™ 11-800 Foam Nitrile Coated Work Gloves
Ansell
HyFlex™ 11-800 Foam Nitrile Coated Work Gloves
Foam nitrile coating enhances grip capabilities which allows workers to easily perform tactile tasks. Gloves are tailored at the base of the pinky finger to better conform to the natural contour of the hand.
Have Questions?
Catalog NumberSize
FIS19-050-067D9
Catalog number FIS19-050-067D
Price (SGD)
208.00
Dozen
Size:
9
Price (SGD)
208.00
Dozen
  • Ansell ZONZ™ Comfort Fit technology for enhanced fit and improved feel
  • New design provides more durability than previous formulations
  • Foam coating increases breathability for user comfort
  • Proprietary washing process efficiently extracts impurities to deliver a cleaner, more comfortable glove
  • Enhanced grip capabilities in dry and slightly oily environments
Specifications
ANSI Abrasion RatingANSI 3
Certifications/ComplianceCE Category II, EN 388:2016+A1:2018 3131A, EN 407:2020 X1XXXX, EN ISO 21420:2020, REACH, OEKO-TEX STANDARD 100, Dermatest, Food Contact
Certifications/Compliance (Europe)CE Category II, EN 388:2016+A1:2018 3131A, EN 407:2020 X1XXXX, EN ISO 21420:2020, REACH
Chemotherapeutic RatedNo
Coated AreaPalm, Fingers
ConstructionKnit
Coating MaterialNitrile Foam
ColorGray, White
Cut Resistance RatingANSI A1
Exterior FinishNo Texture
HandSpecific
Latex FreeYes
MaterialNylon
Product TypeSeamless Knit Gloves
Size9
StyleKnit Wrist
For Use With (Application)Automotive Repairs, Heavy Equipment Operation, Metal Fabrication, and Light Chemical Handling
GenderUnisex
Glove Cuff StyleKnit
Grip DesignSmooth
Liner MaterialNylon
Packaging TypePolybag
Series11-800
Yarn Gauge15
Length (English)8.34 to 10.31 in.
Length (Metric)212 to 262 mm
Product LineHyFlex
Unit SizeDozen