TempDex 720 Nitrile-Coated Thermal Protection Gloves
Actual product may vary
MAPA
TempDex 720 Nitrile-Coated Thermal Protection Gloves
Aramid Fiber knit construction provides cut resistance, while also offering protection from temperatures up to 150°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-046-5549
FIS19-046-5537
FIS19-046-555
also known as FIS 19-046-555
11
Catalog number FIS19-046-554
Price (SGD)
59.00
Pair
Size:
9
Price (SGD)
59.00
Pair
  • Aramid Fiber knit construction offers cut protection
  • Terrycloth liner provides comfort and insulation
  • Nitrile coating enhances grip in moderately oily applications
  • Silicone free to prevent traces or defects on metal sheets and glass
  • Protection from temperatures up to 150°C
Specifications
Coating MaterialNitrile
ColorOrange
MaterialAramid Fiber
Size9
Temperature (English) Max.300°F
Temperature (Metric) Max150°C
For Use With (Application)Ceramic, Hot Material Handling, Steel Manufacturing, and Vulcanization
Glove Cuff StyleKnit
SeriesTempDex 720
Unit SizePair