Acid Suit 96 Coat
Actual product may vary
Neese
Acid Suit 96 Coat
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-046-103D2X-Large
FIS19-046-103CLarge
FIS19-046-147X-Large
Catalog number FIS19-046-103D
Price (SGD)
162.00
Each
Size:
2X-Large
Price (SGD)
162.00
Each
  • Standup collar and snaps for hood
  • Coat style is a minimum of 48 in. long
  • Snap front and storm flap
  • Green PVC/polyester knit
Specifications
Size2X-Large
MaterialPolyester, PVC
ColorGreen
Closure TypeSnap
ConstructionPVC, Polyester Knit
Collar TypeStand-up
ResistanceFlame
For Use With (Application)Chemical, rain, flame
Length (English)48 in.
Length (Metric)121.92 cm
Unit SizeEach