Unispec™ II Safety Glasses
Unispec™ II Safety Glasses
Actual product may vary
Ansell
Unispec™ II Safety Glasses
KleenGuard™ Unispec II Safety Eyewear offers wraparound protection to fit over most prescription eyewear.
Have Questions?
Catalog NumberLens Tint
FIS19-041-232Clear
Catalog number FIS19-041-232
Price (SGD)
350.00
Pack
Add to cart
Lens Tint:
Clear
Price (SGD)
350.00
Pack
Add to cart
  • Low-cost wraparound protection with one-piece clear polycarbonate lens
  • Uncoated clear frame
  • Side shields provide added protection
Specifications
Lens CoatingUncoated
Certifications/ComplianceCE Approved, ANSI Z87.1/CSA Z94.3
Lens TintClear
Product TypeSafety Glasses
MaterialPlastic
DisposableReusable
Lens MaterialPlastic
Frame MaterialPlastic
UV ProtectionUV400
Frame TypeFrameless
Unit SizePack