Scott™ Single-Fold Paper Towels
Actual product may vary
Ansell
Scott™ Single-Fold Paper Towels
For areas that require an economy line of washroom products
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-040-8989.3 x 10.3 in. (23.6 x 26 cm)
Catalog number FIS19-040-898
Price (SGD)
238.00
Case
Dimensions (L x W):
9.3 x 10.3 in. (23.6 x 26 cm)
Price (SGD)
238.00
Case
  • Size: 23.6 × 26cm (9.3 × 10.3 in.)
  • Color: White
  • Recycled fiber content: 60%
  • Post consumer waste: 40%
  • Recyclable container
  • Case dim.: 22.25 × 16.125 × 9.75 in.
Specifications
Product TypeSingle-fold Towel
Length (English)9.3 in.
Length (Metric)23.6 cm
Width (English)10.3 in.
Width (Metric)26 cm
MaterialRecycled Fiber
Certifications/ComplianceEPA
ColorWhite
Dimensions (L x W)9.3 x 10.3 in. (23.6 x 26 cm)
For Use With (Application)Areas in business that require an economy line of washroom products
Ply1-ply
Unit SizeCase