Search Thermo Fisher Scientific
| Catalog Number | Door Style |
|---|---|
| FIS19-039-779 | Manual-close |
Keep acids safe and secure but readily in reach with a SciMat™ Polypropylene Acid Bench Cabinet.
Safe, convenient bench or table top acid storage; Made of rust-free, noncorroding white polypropylene; Welded, liquid-tight interior seams and trough.
Keeps even the most corrosive liquids safely and conveniently within reach on the work surface. Designed to hold up to 6 x2.5L bottles, this durable 16 in. square (41cm) bench cabinet is constructed of rust-free, noncorroding .25 in. thick (0.64cm) white polypropylene, including the hinges. Doors are .5 in. thick white polypropylene with welded, liquid-tight interior seams and a 2 in. (5.1cm) trough to contain spillage.