Spill Containment Drums
Actual product may vary
Eagle
Spill Containment Drums
Approved for use with Packing Groups I, II and III
Have Questions?
Catalog NumberCapacity (English)TypeLoad Bearing Capacity (English)
FIS19-039-66820 gal.Lab Pack Poly Drum, Screw Top125 lb.
Catalog number FIS19-039-668
Price (SGD)
208.00
Each
Capacity (English):
20 gal.
Type:
Lab Pack Poly Drum, Screw Top
Load Bearing Capacity (English):
125 lb.
Price (SGD)
208.00
Each
Lightweight drums for safe storage, transportation, cleanup and containment of containers from 5 to 55 gallons (19 to 208L). Made of chemical- and weather-resistant HDPE with UV inhibitors that incinerate completely. Nestable for ideal storage.
Lab Packs
  • For use with bottles, cans, carboys and 5-gal. pails
  • Yellow (except 1610MB which is blue)
Salvage Drums
  • Meet 3psi pressure test for Salvage Drum Compliance
  • Require “Salvage Drum” labeling (included)
  • Yellow
Overpacks
  • For use with larger containers up to 55 gal.
  • Pass 3psi pressure test
  • Yellow
Specifications
Certifications/ComplianceDOT 49 CFR 173.3, FDA 21 CFR 177.1520
ColorYellow
Capacity (English)20 gal.
Capacity (Metric)76 L
MaterialHDPE
TypeLab Pack Poly Drum, Screw Top
For Use With (Application)Ideal for storage, transportation, clean up and spill containment of larger containers
For Use With (Equipment)Bottles, cans, carboys and 5 gal. pails
Load Bearing Capacity (English)125 lb.
Load Bearing Capacity (Metric)57 kg
ResistantChemical, Weather
Height (English)20.75 in.
Height (Metric)52.7 cm
Weight (English)13 lb.
Weight (Metric)5.85 kg
Unit SizeEach