Haz-Mat Pads
Actual product may vary
Haz-Mat Pads
Haz-Mat Pads
Haz-Mat Pads
New Pig
Haz-Mat Pads
Stop the spread of acids and bases.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Dimensions (L x W)Weight (English)
FIS19-039-15428.16 oz.20 x 15 in.17 lb.
FIS19-033-46812.54 oz.13 x 10 in.8.1 lb.
Catalog number FIS19-039-154
Price (SGD)
353.00
Case
Capacity (English):
28.16 oz.
Dimensions (L x W):
20 x 15 in.
Weight (English):
17 lb.
Price (SGD)
353.00
Case
Stop the spread of acids and bases.
  • Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills
  • Absorb and retain most acids, bases and unknown liquids
  • Can be incinerated after use to reduce waste
Specifications
ColorPink
Capacity (English)28.16 oz.
Capacity (Metric)0.83 L
MaterialPolypropylene
TypeHazmat Absorbent Pad
For Use With (Application)Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives
Weight CapacityHeavyweight
Dimensions (L x W)20 x 15 in.
Length (English)20 in.
Length (Metric)50.8 cm
Width (English)15 in.
Width (Metric)38 cm
Weight (English)17 lb.
Weight (Metric)7.7 kg
Unit SizeCase