Polyethylene Oily Waste Cans
Actual product may vary
Eagle
Polyethylene Oily Waste Cans
Designed for temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)ColorHeight (English)Width (English)
FIS19-038-45414 gal.Yellow21 in.22 in.
FIS19-038-4526 gal.Yellow16 in.16.5 in.
Catalog number FIS19-038-454
Price (SGD)
347.00
Each
Capacity (English):
14 gal.
Color:
Yellow
Height (English):
21 in.
Width (English):
22 in.
Price (SGD)
347.00
Each
Durable, fire-safe cans won't rust or dent and resist most chemical compounds.
  • Cans feature foot treadle that opens cover; self-closing lid
  • Choice of red or high-visibility yellow
Specifications
Capacity (Metric)53 L
Certifications/ComplianceFM, OSHA 29 CFR 1910.108, 20 CFR 1910.106
Depth (English)18 in.
Closure TypeSelf Closing
Depth (Metric)45.7 cm
Capacity (English)14 gal.
ColorYellow
Height (Metric)53 cm
Quantity1 Ea.
Weight (English)16 lb.
Weight (Metric)7.2 kg
Width (Metric)55.8 cm
IncludesFoot lever
For Use With (Application)For temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion
Width (English)22 in.
Height (English)21 in.
MaterialHDPE
Product TypeOily Waste Can
Unit SizeEach