Super Erecta™ SiteSelect™ Posts
Actual product may vary
Super Erecta™ SiteSelect™ Posts
Super Erecta™ SiteSelect™ Posts
Metro
Super Erecta™ SiteSelect™ Posts
For use with Metro Super Erecta, Super Adjustable Super Erecta and Super Erecta Pro wire shelving systems
Have Questions?
Catalog NumberHeight (English)StyleMaterial
FIS19-037470H86.63 in.StationaryStainless Steel (304)
Catalog number FIS19-037470H
Price (SGD)
-
Height (English):
86.63 in.
Style:
Stationary
Material:
Stainless Steel (304)
  • Stationary posts are fitted with adjustable leveling bolts to compensate for uneven surfaces.
  • Posts are available in chrome, type 304 stainless steel, and metroseal 3.
  • Metroseal 3 includes Microban™ antimicrobial product protection and has a 12 year warranty against rust and corrosion.
  • Mobile posts come without leveling bolt assembly for use with stem casters.
  • Grooved at 1 in. (2.5 cm) intervals.
  • Posts are double-grooved every 8 in. (20.3 cm) for easy identification.
Specifications
Product TypeStationary Shelving Post
MaterialStainless Steel (304)
For Use With (Equipment)Metro Super Erecta Wire Shelving System
Height (English)86.63 in.
Height (Metric)220 cm
StyleStationary
FinishMetroseal Epoxy 3
Unit SizeEach