▸
Furniture
▸
Desks and Tables
▸
Laboratory Desks and Workstations
▸
Stainless-Steel Cleanroom Tables
Actual product may vary
Advance Tabco
Stainless-Steel Cleanroom Tables
Featuring flat sides, the Advance Tabco Stainless-Steel Cleanroom Table simplifies alignment and conserves space.
Have Questions?
Catalog Number
Dimensions (L x W x H)
Weight (English)
FIS19-033-543A
48 x 24 x 30 in. (122 x 60.96 x 76.2 cm)
80 lb.
Catalog number
FIS19-033-543A
Price (SGD)
5,246.00
Each
Add to cart
Dimensions (L x W x H):
48 x 24 x 30 in. (122 x 60.96 x 76.2 cm)
Weight (English):
80 lb.
Price (SGD)
5,246.00
Each
Add to cart
For use in ISO Class 5 to 8 cleanrooms (Class 100–100,000)
Constructed from 16-gauge Type 304 series stainless steel
Top with 1.625 in. rolled edge and 2.5 in. solid border dimension on front and back; sides are flat turne down
Solid tops reinforced with welded stainless-steel hat channels
Stainless-steel understructure has center cross bracing to maintain rigidity
1 in. (2.5cm) adjustable stainless-steel bullet feet
Specifications
Dimensions (L x W x H)
48 x 24 x 30 in. (122 x 60.96 x 76.2 cm)
Height (English)
30 in.
Height (Metric)
76.2 cm
Includes
Necessary hardware for easy assembly
Length (English)
48 in.
Length (Metric)
122 cm
Weight (English)
80 lb.
Weight (Metric)
27.66 kg
Width (English)
24 in.
Width (Metric)
60.96 cm
Product Type
Cleanroom Table
Material
Stainless Steel