Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Actual product may vary
NPS Corp.
Spilfyter™ KolorSafe™ Liquid Neutralizers for Bases
Take the guesswork out of neutralizing hazardous spills
Have Questions?
Catalog NumberCapacity (Metric)Packaging Quantity
FIS19-02189218.9 L1 Ea.
Catalog number FIS19-021892
Price (SGD)
697.00
Each
Capacity (Metric):
18.9 L
Packaging Quantity:
1 Ea.
Price (SGD)
697.00
Each
Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.
  • Sodium-free liquid reagent changes color when neutralization is complete
  • Safe for use in laboratories and cleanrooms
Specifications
Capacity (English)5 gal.
Capacity (Metric)18.9 L
TypeBase Neutralizer
FormatLiquid
For Use With (Application)For lab and cleanrooms
IncludesColor change indicator when neutralized
Packaging Quantity1 Ea.
Shelf Life3 Years
Weight (English)50 lb.
Weight (Metric)23 kg
Unit SizeEach