Spilfyter™ Lab and Cleanroom Kollect-a-Kem™ Sorbents
Actual product may vary
NPS Corp.
Spilfyter™ Lab and Cleanroom Kollect-a-Kem™ Sorbents
Available in low-lint and extra-low-lint pads and rolls
Have Questions?
Catalog NumberCapacity (English)Weight CapacityColor
FIS19-0218554 gal.Medium-weightYellow
Catalog number FIS19-021855
Price (SGD)
323.00
Case
Capacity (English):
4 gal.
Weight Capacity:
Medium-weight
Color:
Yellow
Price (SGD)
323.00
Case
Low-Lint Pads and Pillows
  • Use in work areas to keep them safe from spills and chemical overruns
  • Constructed from premium fine fiber Spilfyter Hazmat Pads encased in tear-resistant, Low-Lint Spunbond fabric
Extra-Low-Lint Pads and Rolls
  • Made from Spilfyter Premium Hazmat Sorbents with extra-low-lint heat-pressed material
  • Provide for a 100% lint-free cleanroom
  • Note: It is the end user's responsibility to determine if these sorbents are suitable for use prior to allowing them into the cleanroom environment.
Specifications
ColorYellow
Capacity (English)4 gal.
Capacity (Metric)15.14 L
MaterialPolypropylene
TypeLow-lint Sorbent Pillow
For Use With (Application)Keep work areas safe, free from spills and from liquid or chemical over-run
Weight CapacityMedium-weight
Dimensions (L x W)17 x 8.5 in. (43.18 x 21.5 cm)
Length (English)17 in.
Length (Metric)43.18 cm
Width (English)8.5 in.
Width (Metric)21.5 cm
Weight (English)6 lb.
Weight (Metric)3 kg
Unit SizeCase