Check Order Status
Modesty Curtain
Actual product may vary
Encon
Modesty Curtain
This water-resistant curtain is yellow for high visibility
Have Questions?
Change view
Catalog Number
Product Type
Material
FIS19-021747
Modesty Curtain Assembly
Nylon, PVC
FIS19-086363
Modesty Curtain Hooks
Zinc
2 Options
Catalog number
FIS19-021747
Price (SGD)
-
Product Type:
Modesty Curtain Assembly
Material:
Nylon, PVC
Stainless-steel pipe
ABS plastic blocks
SS rod and support bracket with SS U-bolts to hold assembly in place
PVC nylon coating
Weighted along bottom
178 x 368cm (70 x 145 in.)
Specifications
Color
Yellow
For Use With (Equipment)
Emergency Shower Station
Height (English)
70 in.
Height (Metric)
177.8 cm
Includes
ABS Plastic Blocks, Stainless Steel Rod, Support Bracket with Stainless Steel U-bolts
Material
Nylon, PVC
Product Type
Modesty Curtain Assembly