Ear Plugs
Actual product may vary
Ear Plugs
Contoured shape offers ease of insertion and resistance to backing out of the ear canal. Soft foam offers long-wearing comfort.
Have Questions?
Catalog NumberNoise Reduction Rating (NRR)
FIS19-021-23033 dB
Catalog number FIS19-021-230
Price (SGD)
-
Noise Reduction Rating (NRR):
33 dB
  • Contoured bell shape is easy to insert, resists tendency to back-out of ear canal
  • Polyurethane foam enhances comfort, especially for long-term wear
  • Smooth, soil-resistant closed-cell foam skin prevents dirt build-up
Specifications
ANSI S3.1974Yes
Cord ColorBlue
Cord MaterialVinyl
CSA Z94.2Class AL
SizeUniversal
For Use With (Application)For Work Situations That Demand a High Degree of Comfort, Frequent Changes, or Where Hygiene Presents a Problem for re-Use
High VisibilityYes
Packaging TypePoly bag
Pairs per Package100 Pairs/Pk.
Plug MaterialFoam
Plug ColorOrange
Noise Reduction Rating (NRR)33 dB
CordedCorded
Plug ShapeBell
DisposableSingle-use
Metal DetectableNo
Unit SizePack