ActivArmr™ 42-474 Industrial Gloves
Actual product may vary
ActivArmr™ 42-474 Industrial Gloves
ActivArmr™ 42-474 Industrial Gloves
ActivArmr™ 42-474 Industrial Gloves
Ansell
ActivArmr™ 42-474 Industrial Gloves
Heat-resistant full-length gloves offer complete protection and added comfort
Have Questions?
Catalog NumberSize
FIS19-01985710
Catalog number FIS19-019857
Price (SGD)
-
Size:
10
  • Felt insulation and heat barrier for hot and dry handling (250°C/480°F).
  • Outseam-free two-piece construction for enhanced comfort and flexibility.
  • Bonded nitrile coating provide snag, puncture, abrasion, and cut resistance.
  • Sweat absorbent nonwoven felt liner limits perspiration.
  • Bonded nitrile coating provides additional snag, puncture, cut, and abrasion resistance.
  • Enhanced grip give secure grip in dry or oily conditions.
  • Ideal for use in automotive, chemical, and food processing (indirect contact only).
Specifications
Certifications/Compliance (Europe)EN 420:2003 + A1:2009; Category III
ChlorinatedNo
Coated AreaUncoated
Color CodeGray
ConstructionKnit
Accelerator FreeNo
ApplicationsIndustrial
Coating MaterialNitrile
ColorGray
Cut Resistance RatingANSI A2
Exterior FinishNo Texture
Latex FreeYes
MaterialPolyester
Product TypeHeat-resistant Gloves
Size10
StyleClute
Temperature (English) Max.480°F
Temperature (Metric) Max250°C
EN388 Standard2241B
For Use With (Application)Press Injection Molding, Testing, Welding
GenderUnisex
Glove Cuff ColorGray
Glove Cuff StyleGauntlet
Hem ColorGray
Liner MaterialNon- woven Polyester, Polycotton
LiningLined
Model42-474
Resistant ToHeat
SeriesActivArmr™ 42-474
Thumb StyleStraight
Unit SizeDozen